Gene Transcripts

Gene Symbol
Gria1
Entrez Gene ID
14799
Gene Name
glutamate receptor, ionotropic, AMPA1 (alpha 1)
Gene Aliases
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1
Chromosome Location
Chr. 11: 57011571 - 57330244 on Build GRCm38
UniGene ID
Mm.4920
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113325.2 NP_001106796.1 2 598
NM_001252403.1 NP_001239332.1 2 225
NM_008165.4 NP_032191.2 2 598
GenBank mRNA AB102773.1 - 2 182
AB102774.1 - 2 182
AK049958.1 - 2 408
AK135107.1 - 2 226
BC056397.1 - 2 260 6842391
BC060702.1 - 2 181
JN180918.1 - 249
JN180919.1 - 2 180
X57497.1 - 2 445

View Full Product Details