Gene Transcripts
- Gene Symbol
- Hmox2
- Entrez Gene ID
- 15369
- Gene Name
- heme oxygenase 2
- Gene Aliases
- HO-2, HO2
- Chromosome Location
- Chr. 16: 4726361 - 4766249 on Build GRCm38
- UniGene ID
- Mm.272866
- Species
- Mus musculus
- Gene Symbol
- Cdip1
- Entrez Gene ID
- 66626
- Gene Name
- cell death inducing Trp53 target 1
- Gene Aliases
- 2700048O17Rik, 5730403B10Rik, AU018521, AW742692, CDIP
- Chromosome Location
- Chr. 16: 4765461 - 4790287 on Build GRCm38
- UniGene ID
- Mm.157648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029975.1 |
- |
|
999 |
|
View Full Product Details