Gene Transcripts
- Gene Symbol
- Itm2b
- Entrez Gene ID
- 16432
- Gene Name
- integral membrane protein 2B
- Gene Aliases
- AI256040, Bri, Bri2, Bricd2b, D14Sel6, E25BMM, imBRI2
- Chromosome Location
- Chr. 14: 73362231 - 73385271 on Build GRCm38
- UniGene ID
- Mm.392973
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008410.2 |
NP_032436.1
|
3, 4 |
602 |
|
GenBank mRNA |
AB030203.1 |
- |
3, 4 |
622 |
|
AB030204.1 |
- |
2, 3 |
418 |
|
AK002599.1 |
- |
3, 4 |
565 |
|
AK003224.1 |
- |
3, 4 |
569 |
|
AK003587.1 |
- |
3, 4 |
591 |
|
AK003928.1 |
- |
3, 4 |
590 |
|
AK005125.1 |
- |
3, 4 |
593 |
|
AK076139.1 |
- |
3, 4 |
611 |
|
AK077482.1 |
- |
3, 4 |
604 |
|
AK088270.1 |
- |
3, 4 |
589 |
|
AK150378.1 |
- |
3, 4 |
622 |
|
AK150396.1 |
- |
3, 4 |
621 |
|
AK151714.1 |
- |
3, 4 |
621 |
|
AK151889.1 |
- |
3, 4 |
622 |
|
AK152068.1 |
- |
3, 4 |
610 |
|
AK152516.1 |
- |
3, 4 |
622 |
|
AK152650.1 |
- |
3, 4 |
622 |
|
AK152731.1 |
- |
3, 4 |
593 |
|
AK152983.1 |
- |
3, 4 |
614 |
|
AK153103.1 |
- |
3, 4 |
565 |
|
AK159307.1 |
- |
3, 4 |
565 |
|
AK159340.1 |
- |
3, 4 |
590 |
|
AK159378.1 |
- |
3, 4 |
621 |
|
AK159426.1 |
- |
3, 4 |
590 |
|
AK159619.1 |
- |
3, 4 |
614 |
|
AK159628.1 |
- |
3, 4 |
590 |
|
AK159792.1 |
- |
3, 4 |
590 |
|
AK159962.1 |
- |
3, 4 |
593 |
|
AK160101.1 |
- |
3, 4 |
590 |
|
AK160725.1 |
- |
3, 4 |
621 |
|
AK194069.1 |
- |
|
167 |
|
BC010320.1 |
- |
3, 4 |
605 |
3599190 |
BC021786.1 |
- |
3, 4 |
611 |
4458918 |
U76253.1 |
- |
3, 4 |
602 |
|
View Full Product Details