Gene Transcripts
- Gene Symbol
- Lama3
- Entrez Gene ID
- 16774
- Gene Name
- laminin, alpha 3
- Gene Aliases
- Lama3B, [a]3B
- Chromosome Location
- Chr. 18: 12333723 - 12583013 on Build GRCm38
- UniGene ID
- Mm.42012
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010680.1 |
NP_034810.1
|
11 |
1442 |
|
GenBank mRNA |
U88353.1 |
- |
11 |
1295 |
|
View Full Product Details