Gene Transcripts

Gene Symbol
Olfr51
Entrez Gene ID
18351
Gene Name
olfactory receptor 51
Gene Aliases
ID7, MOR129-3
Chromosome Location
Chr. 11: 51006974 - 51007897 on Build GRCm38
UniGene ID
Mm.246541
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146909.1 NP_667120.1 1 609
GenBank mRNA AF102525.1 - 1 420
BC111814.1 - 1 608

View Full Product Details