Gene Transcripts

Gene Symbol
Pde6g
Entrez Gene ID
18588
Gene Name
phosphodiesterase 6G, cGMP-specific, rod, gamma
Gene Aliases
Pdeg, p3AT
Chromosome Location
Chr. 11: 120447607 - 120453500 on Build GRCm38
UniGene ID
Mm.39200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF190928.1 - 1 20
AK080735.1 - 2 166
BC014723.1 - 2 135
BC082288.1 - 2 123 6490841
Y00746.1 - 2 141

View Full Product Details