Gene Transcripts

Gene Symbol
Ttc36
Entrez Gene ID
192653
Gene Name
tetratricopeptide repeat domain 36
Gene Aliases
Chromosome Location
Chr. 9: 44799400 - 44802951 on Build GRCm38
UniGene ID
Mm.325487
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138951.1 NP_620401.1 2 357
GenBank mRNA AK149384.1 - 2 388
AK165811.1 - 2 255
BC021608.1 - 2 269 5099426
BC021613.1 - 2 363 5102393
BC024432.1 - 2 357 4972715
BC026825.1 - 2 304 4240199

View Full Product Details