Gene Transcripts

Gene Symbol
Dip2c
Entrez Gene ID
208440
Gene Name
DIP2 disco-interacting protein 2 homolog C (Drosophila)
Gene Aliases
2900024P20Rik, 9630044M06, mKIAA0934
Chromosome Location
Chr. 13: 9276503 - 9668928 on Build GRCm38
UniGene ID
Mm.217326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081426.2 NP_001074895.2 8 1403
GenBank mRNA BC137787.1 - 8 995
BC144909.1 - 9 1163 9053457
BC144910.1 - 8 995 9053458

View Full Product Details