Gene Transcripts

Gene Symbol
Metrnl
Entrez Gene ID
210029
Gene Name
meteorin, glial cell differentiation regulator-like
Gene Aliases
9430048M07Rik, BC019776
Chromosome Location
Chr. 11: 121702427 - 121717389 on Build GRCm38
UniGene ID
Mm.153566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144797.3 NP_659046.1 3, 4 805
GenBank mRNA AK034855.1 - 3, 4 806
AK085880.1 - 3, 4 807
AK170549.1 - 2, 3 420
AK170800.1 - 3, 4 807
AK171200.1 - 3, 4 551
BC019776.1 - 3, 4 760 3672224
BC024445.1 - 3, 4 820 4974030
BC024497.1 - 808 4982563
BC026646.1 - 3, 4 799 4987595

View Full Product Details