Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011594.3 | NP_035724.2 | 5 | 893 | |
GenBank mRNA | AK032818.1 | - | 5 | 807 | |
AK081522.1 | - | 5 | 990 | ||
AK144236.1 | - | 5 | 807 | ||
AK192526.1 | - | 3 | 264 | ||
BC049126.1 | - | 5 | 790 | 6821553 | |
M82858.1 | - | 5 | 788 | ||
X62622.1 | - | 5 | 798 |