Gene Transcripts

Gene Symbol
Tuft1
Entrez Gene ID
22156
Gene Name
tuftelin 1
Gene Aliases
Chromosome Location
Chr. 3: 94612753 - 94658872 on Build GRCm38
UniGene ID
Mm.10214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293728.1 NP_001280657.1 7 646
NM_011656.3 NP_035786.1 8 721
GenBank mRNA AF002860.1 - 8 615
AF047704.1 - 8 655
AK132488.1 - 8 653
AK171705.1 - 7 611
BC019213.1 - 8 652 5064658

View Full Product Details