Gene Transcripts

Gene Symbol
Pla2g16
Entrez Gene ID
225845
Gene Name
phospholipase A2, group XVI
Gene Aliases
C78643, HRSL3, Hrasls3, Hrev107, MLP-3
Chromosome Location
Chr. 19: 7557434 - 7588545 on Build GRCm38
UniGene ID
Mm.274810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139269.2 NP_644675.2 4 374
GenBank mRNA AB298806.1 - 2 219
AK039703.1 - 4 587
AK042007.1 - 4 327
AK052657.1 - 4 353
AK163505.1 - 4 375
AX775891.1 - 4 356

View Full Product Details