Gene Transcripts
- Gene Symbol
- Lzts2
- Entrez Gene ID
- 226154
- Gene Name
- leucine zipper, putative tumor suppressor 2
- Gene Aliases
- BC014695, LAPSER1, mKIAA1813
- Chromosome Location
- Chr. 19: 45015176 - 45027115 on Build GRCm38
- UniGene ID
- Mm.28204
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC014695.1 |
- |
2 |
1007 |
|
BC092263.1 |
- |
3 |
1138 |
2654420 |
View Full Product Details