Gene Transcripts

Gene Symbol
Sowaha
Entrez Gene ID
237761
Gene Name
sosondowah ankyrin repeat domain family member A
Gene Aliases
A830006N08Rik, Ankrd43
Chromosome Location
Chr. 11: 53476578 - 53480195 on Build GRCm38
UniGene ID
Mm.145208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183173.2 NP_898996.2 1 1371
GenBank mRNA AK043538.1 - 1 1258
BC117863.1 - 1 709 40095300

View Full Product Details