Gene Transcripts

Gene Symbol
Banf1
Entrez Gene ID
23825
Gene Name
barrier to autointegration factor 1
Gene Aliases
Baf, Bcrp1, L2bp1/Baf
Chromosome Location
Chr. 19: 5364633 - 5366876 on Build GRCm38
UniGene ID
Mm.358649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038231.2 NP_001033320.1 2 198
NM_001286608.1 NP_001273537.1 2 540
NM_011793.3 NP_035923.1 2 406
GenBank mRNA AB025349.1 - 2 138
AF037080.1 - 1 81
AK003025.1 - 2 182
AK004212.1 - 2 165
AK010257.1 - 2 184
AK011079.1 - 2 187
AK011100.1 - 2 185
AK012588.1 - 2 168
AK088896.1 - 2 162
AK168039.1 - 2 188
AK189231.1 - 1 99
BC003709.1 - 2 175 3599828
BY111854.1 - 2 271
CK780850.1 - 2 170

View Full Product Details