Gene Transcripts

Gene Symbol
C1ql1
Entrez Gene ID
23829
Gene Name
complement component 1, q subcomponent-like 1
Gene Aliases
C1qrf, CRF, CTRP14, gliacolin
Chromosome Location
Chr. 11: 102939264 - 102946461 on Build GRCm38
UniGene ID
Mm.57154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011795.2 NP_035925.2 1 399
GenBank mRNA AF095155.1 - 1 399
BC118980.2 - 1 399 40100122

View Full Product Details