Gene Transcripts

Gene Symbol
Syne2
Entrez Gene ID
319565
Gene Name
spectrin repeat containing, nuclear envelope 2
Gene Aliases
6820443O06Rik, AW546258, D12Ertd777e, NUA, Nesp2g, mKIAA1011
Chromosome Location
Chr. 12: 75818318 - 76110928 on Build GRCm38
UniGene ID
Mm.26652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005510.2 NP_001005510.2 17 2142
GenBank mRNA AK076294.1 - 2 237
AK078542.1 - 17 2143
AK145252.1 - 17 2144

View Full Product Details