Gene Transcripts

Gene Symbol
Fam71f1
Entrez Gene ID
330277
Gene Name
family with sequence similarity 71, member F1
Gene Aliases
GARI-L1, Gm764, NYD-SP18
Chromosome Location
Chr. 6: 29313482 - 29336022 on Build GRCm38
UniGene ID
Mm.87624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289663.1 NP_001276592.1 3 511
NM_001289664.1 NP_001276593.1 3 511
NM_001289665.1 NP_001276594.1 3 506
NM_207258.3 NP_997141.2 3 511
GenBank mRNA AB473725.1 - 3 452
AK133908.1 - 3 512
BC147513.1 - 3 494 9056505
BC172074.1 - 3 494 9054360
BC172085.1 - 3 489
BC172086.1 - 3 494 9054372

View Full Product Details