Gene Transcripts
- Gene Symbol
- Rtl1
- Entrez Gene ID
- 353326
- Gene Name
- retrotransposon-like 1
- Gene Aliases
- 6430411K18Rik, Mar, Mart1, Mor1, Peg11
- Chromosome Location
- Chr. 12: 109590169 - 109595403 on Build GRCm38
- UniGene ID
- Mm.23062
- Species
- Mus musculus
- Gene Symbol
- 6430411K18Rik
- Entrez Gene ID
- 76880
- Gene Name
- RIKEN cDNA 6430411K18 gen
- Gene Aliases
- PEG11as
- Chromosome Location
- Chr. 12: 109591537 - 109592851 on Build GRCm38
- UniGene ID
- Mm.490311
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018276.1 |
- |
1 |
187 |
|
EU434921.1 |
- |
|
1156 |
|
View Full Product Details