Gene Transcripts

Gene Symbol
Nanos2
Entrez Gene ID
378430
Gene Name
nanos homolog 2 (Drosophila)
Gene Aliases
nos2
Chromosome Location
Chr. 7: 18987524 - 18988962 on Build GRCm38
UniGene ID
Mm.277153
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194064.2 NP_918953.2 1 429
GenBank mRNA AB095972.1 - 1 429
JQ936992.1 - 1 359

View Full Product Details