Gene Transcripts

Gene Symbol
Hnrnph3
Entrez Gene ID
432467
Gene Name
heterogeneous nuclear ribonucleoprotein H3
Gene Aliases
AA693301, AI666703, Hnrph3
Chromosome Location
Chr. 10: 63014650 - 63023889 on Build GRCm38
UniGene ID
Mm.274784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079824.1 NP_001073293.1 2 172
GenBank mRNA BC049652.1 - 2 170
BC147496.1 - 2 153
BQ443117.1 - 1 13
BU946493.1 - 2 173

Gene Symbol
Aff3
Entrez Gene ID
16764
Gene Name
AF4/FMR2 family, member 3
Gene Aliases
3222402O04Rik, A730046J16, LAF-4, Laf4
Chromosome Location
Chr. 1: 38175991 - 38627837 on Build GRCm38
UniGene ID
Mm.336679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC094538.1 - 13

View Full Product Details