Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015811.2 | NP_056626.2 | 2 | 251 | |
GenBank mRNA | AF215667.1 | - | 2 | 182 | |
AK089293.1 | - | 2 | 207 | ||
AK138688.1 | - | 2 | 207 | ||
AK154394.1 | - | 207 | |||
AK154841.1 | - | 544 | |||
AK170366.1 | - | 2 | 203 | ||
AK170957.1 | - | 2 | 207 | ||
AK171306.1 | - | 548 | |||
BC028634.1 | - | 2 | 182 | 1328269 | |
BC052684.1 | - | 1 | 526 | ||
BU695313.1 | - | 2 | 273 |