Gene Transcripts

Gene Symbol
Ppap2c
Entrez Gene ID
50784
Gene Name
phosphatidic acid phosphatase type 2C
Gene Aliases
Lpp2, PAP2-G, PAP2-gamma, PAP2c
Chromosome Location
Chr. 10: 79526424 - 79533787 on Build GRCm38
UniGene ID
Mm.28873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK005452.1 - 2 223
AK049581.1 - 2 266
AK179023.1 - 1 27
BC010332.1 - 2 244 2654393
BC016635.1 - 2 194
BC085104.1 - 2 182
BY344892.1 - 2 269
CA540403.1 - 2 189


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF123611.1 - 258 Chr. - - [-]

View Full Product Details