Gene Transcripts
- Gene Symbol
- Ccnc
- Entrez Gene ID
- 51813
- Gene Name
- cyclin C
- Gene Aliases
- AI451004, AU020987, CG1C
- Chromosome Location
- Chr. 4: 21727701 - 21759922 on Build GRCm38
- UniGene ID
- Mm.278584
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122982.2 |
NP_001116454.1
|
4, 5 |
485 |
|
NM_001290420.1 |
NP_001277349.1
|
4, 5 |
485 |
|
NM_001290422.1 |
NP_001277351.1
|
4, 5 |
485 |
|
NM_016746.4 |
NP_058026.2
|
4, 5 |
485 |
|
GenBank mRNA |
AK009615.1 |
- |
4, 5 |
430 |
|
AK014079.1 |
- |
3, 4 |
334 |
|
AK035762.1 |
- |
4, 5 |
446 |
|
AK037957.1 |
- |
4, 5 |
486 |
|
AK135472.1 |
- |
4, 5 |
481 |
|
AK153035.1 |
- |
4, 5 |
418 |
|
BB631264.1 |
- |
4, 5 |
483 |
|
BC003344.1 |
- |
4, 5 |
418 |
3257730 |
BC029211.1 |
- |
4, 5 |
431 |
4526925 |
BC062376.1 |
- |
4, 5 |
400 |
|
BC087544.1 |
- |
4, 5 |
417 |
|
BC100396.1 |
- |
4, 5 |
459 |
|
BC120649.1 |
- |
4, 5 |
412 |
40129572 |
BC120677.1 |
- |
4, 5 |
412 |
40129600 |
U62638.1 |
- |
4, 5 |
460 |
|
View Full Product Details