Gene Transcripts

Gene Symbol
Bysl
Entrez Gene ID
53414
Gene Name
bystin-like
Gene Aliases
Bys, Enp1
Chromosome Location
Chr. 17: 47599331 - 47611492 on Build GRCm38
UniGene ID
Mm.472101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016859.3 NP_058555.3 2 706
GenBank mRNA AK019239.1 - 2 441
AK032993.1 - 2 438
AK049195.1 - 2 699
AK051704.1 - 2 696
AK083033.1 - 2 693
AK143027.1 - 2 707
AK168301.1 - 2 444
AK183817.1 - 120
BC017530.1 - 2 433 4925307

View Full Product Details