Gene Transcripts

Gene Symbol
Psors1c2
Entrez Gene ID
57390
Gene Name
psoriasis susceptibility 1 candidate 2 (human)
Gene Aliases
AI448740, Pcg, SPR1
Chromosome Location
Chr. 17: 35532207 - 35534647 on Build GRCm38
UniGene ID
Mm.34201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020576.2 NP_065601.2 2 247
GenBank mRNA AF159091.1 - 2 179
AF484421.1 - 2 206
AK137759.1 - 2 248
BC110561.2 - 2 175 40049760

View Full Product Details