Gene Transcripts

Gene Symbol
Tas2r137
Entrez Gene ID
574417
Gene Name
taste receptor, type 2, member 137
Gene Aliases
T2R37, Tas2r3, mt2r41
Chromosome Location
Chr. 6: 40491238 - 40492239 on Build GRCm38
UniGene ID
Mm.386354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025385.1 NP_001020556.1 1 291

View Full Product Details