Gene Transcripts

Gene Symbol
Ndufa13
Entrez Gene ID
67184
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13
Gene Aliases
2700054G14Rik, AU022060, CDA016, CGI-39, GRIM-19, Grim19
Chromosome Location
Chr. 8: 69887788 - 69902558 on Build GRCm38
UniGene ID
Mm.21162
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023312.2 NP_075801.1 5 1141
GenBank mRNA AF286698.1 - 4 376
AK002999.1 - 4 380
AK003012.1 - 4 394
AK010517.1 - 4 380
AK012422.1 - 4 380
BC034899.1 - 5 1141 1447015
BC037149.1 - 4 384 5149812
BC055435.1 - 4 390 4219489

View Full Product Details