Gene Transcripts

Gene Symbol
Mtfp1
Entrez Gene ID
67900
Gene Name
mitochondrial fission process 1
Gene Aliases
1700020C11Rik, 2610507A21Rik, AV005788, Mtp18
Chromosome Location
Chr. 11: 4091481 - 4095431 on Build GRCm38
UniGene ID
Mm.41719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026443.4 NP_080719.2 2 268
GenBank mRNA AK006148.1 - 2 189
AK010971.1 - 2 225
AK012058.1 - 2 235
AK078381.1 - 2 269
AK167990.1 - 2 135
BC019205.1 - 2 278
BC058737.1 - 2 218 5400435

View Full Product Details