Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167921.1 | NP_001161393.1 | 8 | 893 | |
NM_001167922.1 | NP_001161394.1 | 8 | 995 | ||
NM_026584.3 | NP_080860.3 | 8 | 881 | ||
GenBank mRNA | AK007492.1 | - | 7 | 838 | |
AK021183.1 | - | 8 | 846 | ||
AK088342.1 | - | 8 | 906 | ||
AK146803.1 | - | 8 | 867 | ||
AK150415.1 | - | 8 | 839 | ||
AK153370.1 | - | 8 | 839 | ||
AK158783.1 | - | 8 | 975 | ||
BC020016.1 | - | 8 | 861 | 3501113 | |
DV042794.1 | - | 3 | 203 |