Gene Transcripts
- Gene Symbol
- Sympk
- Entrez Gene ID
- 68188
- Gene Name
- symplekin
- Gene Aliases
- 1500016F02Rik, 4632415H16Rik, AA125406, AI449890, SPK, SYM
- Chromosome Location
- Chr. 7: 19024352 - 19054623 on Build GRCm38
- UniGene ID
- Mm.130902
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026605.2 |
NP_080881.2
|
18, 19 |
2647 |
|
GenBank mRNA |
AK019497.1 |
- |
13, 14 |
2123 |
|
AK146894.1 |
- |
18, 19 |
2628 |
|
AK155093.1 |
- |
18, 19 |
2648 |
|
AK155375.1 |
- |
18, 19 |
2602 |
|
AK165591.1 |
- |
18, 19 |
2626 |
|
AK168541.1 |
- |
18, 19 |
2628 |
|
AK168757.1 |
- |
18, 19 |
2628 |
|
AK172040.1 |
- |
19, 20 |
2676 |
|
AK183486.1 |
- |
1, 2 |
162 |
|
AK194007.1 |
- |
1, 2 |
38 |
|
AK211092.1 |
- |
1, 2 |
38 |
|
AK215684.1 |
- |
1, 2 |
38 |
|
BC049852.1 |
- |
18, 19 |
2592 |
4163760 |
View Full Product Details