Gene Transcripts

Gene Symbol
Krt42
Entrez Gene ID
68239
Gene Name
keratin 42
Gene Aliases
2410039E07Rik, K17n, Ka22, ecat6
Chromosome Location
Chr. 11: 100262882 - 100272565 on Build GRCm38
UniGene ID
Mm.158190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212483.2 NP_997648.2 2 492
GenBank mRNA AY459292.1 - 2 492

View Full Product Details