Gene Transcripts

Gene Symbol
Ecscr
Entrez Gene ID
68545
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
1110006O17Rik, ARIA
Chromosome Location
Chr. 18: 35713086 - 35722725 on Build GRCm38
UniGene ID
Mm.329362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033141.1 NP_001028313.1 3 297
GenBank mRNA AK157485.1 - 3 297
EU025067.1 - 3 233

View Full Product Details