Gene Transcripts

Gene Symbol
Flnc
Entrez Gene ID
68794
Gene Name
filamin C, gamma
Gene Aliases
1110055E19Rik, ABP-280, ABPL, Fln2
Chromosome Location
Chr. 6: 29433127 - 29461888 on Build GRCm38
UniGene ID
Mm.39046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081185.1 NP_001074654.1 42 7381
GenBank mRNA AK135523.1 - 7 1221
BC039885.1 - 7 1249
BC052186.1 - 15 2330
BC060276.1 - 21 3442
BC151097.1 - 42 7186 9088010
BC158128.1 - 41 7087 9007396
BC172159.1 - 41 7087

View Full Product Details