Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168516.1 | NP_001161988.1 | 2 | 95 | |
NM_001168517.1 | NP_001161989.1 | 2 | 142 | ||
NM_027476.3 | NP_081752.2 | 2 | 388 | ||
GenBank mRNA | AK017737.1 | - | 2 | 353 | |
AK028164.1 | - | 2 | 351 | ||
AK043923.1 | - | 2 | 345 | ||
AK169337.1 | - | 2 | 347 | ||
AK170233.1 | - | 2 | 372 | ||
AK170706.1 | - | 2 | 372 | ||
AK171568.1 | - | 2 | 372 | ||
AK172151.1 | - | 2 | 142 | ||
BC056621.1 | - | 2 | 110 | ||
BC071194.1 | - | 2 | 329 | ||
CA559443.1 | - | 2 | 95 | ||
CD354313.1 | - | 2 | 51 |