Gene Transcripts

Gene Symbol
Mmrn1
Entrez Gene ID
70945
Gene Name
multimerin 1
Gene Aliases
4921530G03Rik, Emilin4, Mmrn
Chromosome Location
Chr. 6: 60944317 - 60989378 on Build GRCm38
UniGene ID
Mm.389770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163507.1 NP_001156979.1 6 2005
NM_027613.1 NP_081889.1 6 2005
GenBank mRNA AK036724.1 - 6 1843
AK137556.1 - 6 1844
BC137623.1 - 6 1805 8860642
BC144789.1 - 6 1805 9053337

View Full Product Details