Gene Transcripts

Gene Symbol
Tmem30c
Entrez Gene ID
71027
Gene Name
transmembrane protein 30C
Gene Aliases
4933401B01Rik, 4933409A18Rik, Cdc50c
Chromosome Location
Chr. 16: 57266138 - 57301893 on Build GRCm38
UniGene ID
Mm.158563
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027651.1 NP_081927.1 7 894
GenBank mRNA AK016597.1 - 7 908
AK161475.1 - 6 2758

View Full Product Details