Gene Transcripts

Gene Symbol
Epg5
Entrez Gene ID
100502841
Gene Name
ectopic P-granules autophagy protein 5 homolog (C. elegans)
Gene Aliases
4732475F16, 5430411K18Rik, AI661957, AW456499, mKIAA1632
Chromosome Location
Chr. 18: 77938408 - 78037323 on Build GRCm38
UniGene ID
Mm.297782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195633.1 NP_001182562.1 7 1633
GenBank mRNA AK028969.1 - 3 208

View Full Product Details