Gene Transcripts

Gene Symbol
Fbxl22
Entrez Gene ID
74165
Gene Name
F-box and leucine-rich repeat protein 22
Gene Aliases
1110004B15Rik
Chromosome Location
Chr. 9: 66508459 - 66514593 on Build GRCm38
UniGene ID
Mm.297998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175206.4 NP_780415.1 1 246
GenBank mRNA AK052175.1 - 1 232
AK084845.1 - 1 162
AK085980.1 - 1 189
BC100535.1 - 1 242 30138645

View Full Product Details