Gene Transcripts

Gene Symbol
Klc4
Entrez Gene ID
74764
Gene Name
kinesin light chain 4
Gene Aliases
1200014P03Rik, AA408085, AW146303, Knsl8
Chromosome Location
Chr. 17: 46630624 - 46646167 on Build GRCm38
UniGene ID
Mm.279599
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029091.2 NP_083367.1 15 1839
GenBank mRNA AK004774.1 - 15 1840
AK170793.1 - 15 1823
AK219585.1 - 11

View Full Product Details