Gene Transcripts

Gene Symbol
Izumo2
Entrez Gene ID
75510
Gene Name
IZUMO family member 2
Gene Aliases
1700023D19Rik
Chromosome Location
Chr. 7: 44708698 - 44719845 on Build GRCm38
UniGene ID
Mm.273301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029317.1 NP_083593.1 3 374
GenBank mRNA AK006268.1 - 3 348
BC048526.1 - 3 330 6772588

View Full Product Details