Gene Transcripts

Gene Symbol
Celf3
Entrez Gene ID
78784
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
4930415M08Rik, BRUNOL1, CAGH4, ERDA4, Tnrc4
Chromosome Location
Chr. 3: 94478285 - 94492199 on Build GRCm38
UniGene ID
Mm.44292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289613.1 NP_001276542.1 4 851
NM_001289616.1 NP_001276545.1 5 333
NM_001289620.1 NP_001276549.1 5 333
NM_172434.3 NP_766022.1 4 848
GenBank mRNA AK019578.1 - 2 560
AK030259.1 - 3 159
AK044069.1 - 4 281
AK044506.1 - 5 297
AK087536.1 - 5 293
AY165052.1 - 4 566
BC031740.1 - 1 12
BC057553.1 - 4 906 6401356
BY089288.1 - 5 337

View Full Product Details