Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030262.3 | NP_084538.2 | 3 | 422 | |
GenBank mRNA | AF455270.1 | - | 3 | 412 | |
AK132373.1 | - | 3 | 419 | ||
AK147414.1 | - | 2 | 2017 | ||
AK148244.1 | - | 3 | 421 | ||
BC018194.1 | - | 3 | 420 | ||
BC147077.1 | - | 3 | 489 | 9056064 | |
BC147078.1 | - | 3 | 489 | 9056063 | |
CO814561.1 | - | 422 |