Gene Transcripts

Gene Symbol
Syt12
Entrez Gene ID
191595
Gene Name
synaptotagmin XII
Gene Aliases
Srg1
Chromosome Location
Chr. 1: 219688969 - 219720292 on Build Rnor_6.0
UniGene ID
Rn.10682
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138835.1 NP_620190.1 5 916
XM_008760071.1 XP_008758293.1 5 844
XM_008760072.1 XP_008758294.1 6 890
GenBank mRNA U71294.1 - 5 916

View Full Product Details