Gene Transcripts

Gene Symbol
Adrbk1
Entrez Gene ID
25238
Gene Name
adrenergic, beta, receptor kinase 1
Gene Aliases
BARK1, GRK-2, Grk2
Chromosome Location
Chr. 1: 219536220 - 219544329 on Build Rnor_6.0
UniGene ID
Rn.13010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012776.1 NP_036908.1 1, 2 331
GenBank mRNA M87854.1 - 1, 2 331

View Full Product Details