Gene Transcripts

Gene Symbol
Kctd1
Entrez Gene ID
291772
Gene Name
potassium channel tetramerization domain containing 1
Gene Aliases
Vad, Vad5
Chromosome Location
Chr. 18: 6374778 - 6474990 on Build Rnor_6.0
UniGene ID
Rn.27416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100516.1 NP_001093986.1 3 2064
GenBank mRNA CK473534.1 - 2 164

View Full Product Details