Gene Transcripts

Gene Symbol
Vom2r11
Entrez Gene ID
292417
Gene Name
vomeronasal 2 receptor, 11
Gene Aliases
RGD1563778
Chromosome Location
Chr. 1: 86604426 - 86624214 on Build Rnor_6.0
UniGene ID
Rn.217940
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099470.1 NP_001092940.1 3 728

View Full Product Details