Gene Transcripts

Gene Symbol
Pnma3
Entrez Gene ID
293840
Gene Name
paraneoplastic Ma antigen 3
Gene Aliases
RGD1563752
Chromosome Location
Chr. X: 153063307 - 153067500 on Build Rnor_6.0
UniGene ID
Rn.218329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106342.2 NP_001099812.1 1 409
XM_006229513.2 XP_006229575.1 2 857
XM_006229514.2 XP_006229576.1 2 857

View Full Product Details