Gene Transcripts

Gene Symbol
Lactb
Entrez Gene ID
300803
Gene Name
lactamase, beta
Gene Aliases
Chromosome Location
Chr. 8: 72750174 - 72766307 on Build Rnor_6.0
UniGene ID
Rn.73451
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106833.1 NP_001100303.1 4 738
XM_008766242.1 XP_008764464.1 4 721

View Full Product Details