Gene Transcripts

Gene Symbol
Hspa12a
Entrez Gene ID
307997
Gene Name
heat shock protein 12A
Gene Aliases
Chromosome Location
Chr. 1: 279946811 - 280015358 on Build Rnor_6.0
UniGene ID
Rn.18598
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107445.1 NP_001100915.1 12 2629

View Full Product Details